Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-(4-tert-Butylphenyl)di(2-pyrrolyl)methane, 99%

$167.78 - $674.20

Chemical Identifiers

Chemical Name or Material alpha-(4-tert-Butylphenyl)di(2-pyrrolyl)methane
CAS 167482-98-6
Molecular Formula C19H22N2
Formula Weight 278.39
MDL Number MFCD03454220
Synonym 2,2'-(4-tert-Butylphenyl)methylenebispyrrole; 5-(4-tert-Butylphenyl)dipyrromethane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5106603

 
alfa aesar™
H5106603
1g Each for $167.78

AAH5106606

 
alfa aesar™
H5106606
5g Each for $674.20
Description & Specifications

Specifications

Formula Weight 278.39
Health Hazard 1 Warning
MDL Number MFCD03454220
CAS 167482-98-6
Chemical Name or Material alpha-(4-tert-Butylphenyl)di(2-pyrrolyl)methane
Assay 99%
Molecular Formula C19H22N2
Synonym 2,2'-(4-tert-Butylphenyl)methylenebispyrrole; 5-(4-tert-Butylphenyl)dipyrromethane
Melting Point 158° to 162°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.