Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-(4-Morpholinyl)phenylacetonitrile, 98+%

Chemical Identifiers

Chemical Name or Material alpha-(4-Morpholinyl)phenylacetonitrile
CAS 15190-10-0
Molecular Formula C12H14N2O
Formula Weight 202.26
MDL Number MFCD00014619
UN Number UN3439
Beilstein 181704
Synonym alpha-(4-Morpholino)phenylacetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1237014

  SDS
alfa aesar™
L1237014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1237006

  SDS
alfa aesar™
L1237006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 202.26
CAS 15190-10-0
Chemical Name or Material alpha-(4-Morpholinyl)phenylacetonitrile
Assay ≥98%
Molecular Formula C12H14N2O
MDL Number MFCD00014619
UN Number UN3439
Beilstein 181704
Health Hazard 1 Danger
Melting Point 67° to 71°C
Health Hazard 2 GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Synonym alpha-(4-Morpholino)phenylacetonitrile
Quantity 25g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.