missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-(4-Morpholinyl)phenylacetonitrile, 98+%

Manufacturer:  Alfa Aesar™ L1237006

 View more versions of this product

Catalog No. AAL1237006



Specifications

Specifications

alpha-(4-Morpholinyl)phenylacetonitrile
5g
C12H14N2O
UN3439
2-morpholino-2-phenylacetonitrile, morpholin-4-yl phenyl acetonitrile, 2-morpholin-4-yl-2-phenylacetonitrile, alpha-4-morpholino phenylacetonitrile, morpholino phenyl acetonitrile, 4-morpholineacetonitrile, .alpha.-phenyl, morpholin-4-yl-phenyl-acetonitrile, 2-morpholin-4-yl-2-phenylethanenitrile, alpha-4-morpholinyl phenylacetonitrile, maybridge1_000080
C1COCCN1C(C#N)C2=CC=CC=C2
202.257
202.26
67°C to 71°C
15190-10-0
MFCD00014619
181704
MWZPYQLYZXTCLZ-UHFFFAOYSA-N
2-morpholin-4-yl-2-phenylacetonitrile
85812
≥98%

Chemical Identifiers

15190-10-0
202.257
MWZPYQLYZXTCLZ-UHFFFAOYSA-N
85812
C1COCCN1C(C#N)C2=CC=CC=C2
C12H14N2O
MFCD00014619
2-morpholino-2-phenylacetonitrile, morpholin-4-yl phenyl acetonitrile, 2-morpholin-4-yl-2-phenylacetonitrile, alpha-4-morpholino phenylacetonitrile, morpholino phenyl acetonitrile, 4-morpholineacetonitrile, .alpha.-phenyl, morpholin-4-yl-phenyl-acetonitrile, 2-morpholin-4-yl-2-phenylethanenitrile, alpha-4-morpholinyl phenylacetonitrile, maybridge1_000080
2-morpholin-4-yl-2-phenylacetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents