Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha,3-Dimethylstyrene

Chemical Identifiers

Chemical Name or Material alpha,3-Dimethylstyrene
CAS 1124-20-5
Molecular Formula C10H12
Formula Weight 132.21
MDL Number MFCD08060972
UN Number UN3295
Synonym 1-Isopropenyl-3-methylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5076003

 
alfa aesar™
H5076003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH50760MD

 
alfa aesar™
H50760MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 132.21
Chemical Name or Material alpha,3-Dimethylstyrene
Odor Characteristic
Molecular Formula C10H12
MDL Number MFCD08060972
CAS 1124-20-5
UN Number UN3295
Synonym 1-Isopropenyl-3-methylbenzene
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C