missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allyltriphenylphosphonium chloride, 97%, ACROS Organics™

$48.19 - $48.19

Chemical Identifiers

CAS 18480-23-4
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C21 H20 Cl P
Linear Formula H2CCHCH2P(C6H5)3Cl
Formula Weight 338.82
MDL Number MFCD00031542
Chemical Name or Material Allyltriphenylphosphonium chloride
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC352360050 SDS Thermo Fisher Scientific
35236-0050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC352360250 SDS Thermo Fisher Scientific
35236-0250
Glass bottle 25g Each for $48.19
Specifications

Description

For C-C Bond Formation (Olefination)
Description & Specifications

Chemical Identifiers

18480-23-4
100.0
C21 H20 Cl P
338.82
Allyltriphenylphosphonium chloride
96.0
97%
H2CCHCH2P(C6H5)3Cl
MFCD00031542
≥96.0%

Specifications

18480-23-4
100.0
Glass bottle
97%
H2CCHCH2P(C6H5)3Cl
MFCD00031542
226° to 230°C
Allyltriphenylphosphonium chloride
96.0
Authentic
Crystalline Powder
C21 H20 Cl P
338.82
Beige
5g
≥96.0%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 242-368-7