Please login to your online account to display your discounted pricing

Allyltriphenylphosphonium Bromide 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  103040050

 View more versions of this product

Catalog No. AC103040050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 383.26
Physical Form Powder
Quantity 5g
Color White
EINECS Number 216-332-6
RTECS Number TA1843000
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Molecular Formula C21H20BrP
Assay Percent Range 99%
CAS Min % 98.5
CAS 1560-54-9
Name Note 99%
Chemical Name or Material Allyltriphenylphosphonium bromide
Linear Formula H2C=CHCH2P(C6H5)3Br
MDL Number MFCD00011808
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 MOISTURE SENSITIVE

Specifications

Chemical Name or Material Allyltriphenylphosphonium bromide
Name Note 99%
CAS 1560-54-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C21H20BrP
Linear Formula H2C=CHCH2P(C6H5)3Br
Formula Weight 383.26
MDL Number MFCD00011808
Accessories

MOISTURE SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents