missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Allyltrimethylsilane, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 196992500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Allyltrimethylsilane | |
0.7190g/mL | |
7°C | |
99% | |
C6H14Si | |
H2C=CHCH2Si(CH3)3 | |
10,6; 11,16; 12,23; 13,11; 14,18; 15,8 | |
15,29 | |
Solubility in water: insoluble | |
C[Si](C)(C)CC=C | |
114.27 | |
114.27 |
762-72-1 | |
85.0°C | |
Authentic | |
Glass bottle | |
1.4060 to 1.4080 | |
250mL | |
0.719 | |
allyltrimethylsilane, silane, trimethyl-2-propenyl, trimethylallylsilane, silane, allyltrimethyl, allyl trimethylsilane, unii-8b84c337vf, ccris 2649, allyl trimethyl silane, 3-trimethylsilyl propene, allytrimethylsilane | |
HYWCXWRMUZYRPH-UHFFFAOYSA-N | |
trimethyl(prop-2-enyl)silane | |
69808 | |
99% |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 212-104-5
RUO â Research Use Only