Please login to your online account to display your discounted pricing

Allyltrimethylsilane, 97%, ACROS Organics™

$64.78 - $251.52

Chemical Identifiers

Chemical Name or Material Allyltrimethylsilane
Name Note 97%
CAS 762-72-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H14Si
Linear Formula HC=CHCHSi(CH3)3
Formula Weight 114.27
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC349990100

  SDS
thermo fisher scientific
349990100
Glass bottle 10mL Each for $64.78

AC349990500

  SDS
thermo fisher scientific
349990500
Glass bottle 50mL Each for $251.52
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 114.27
Physical Form Liquid
Quantity 10mL
Color Clear colorless
Merck Index 15, 290
TSCA TSCA
EINECS Number 212-104-5
Fieser 10,6; 11,16; 12,23; 13,11; 14,18; 15,8
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C6H14Si
CAS 762-72-1
Name Note 97%
Linear Formula HC=CHCHSi(CH3)3
MDL Number MFCD00008635
Chemical Name or Material Allyltrimethylsilane
Assay Percent Range 97%
Refractive Index 1.4060 to 1.4080 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.