Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyltriethylammonium bromide, 98+%

$57.57 - $845.30

Chemical Identifiers

Chemical Name or Material Allyltriethylammonium bromide
CAS 29443-23-0
Molecular Formula CH2=CHCH2N(CH2CH3)3Br
Formula Weight 222.18
MDL Number MFCD00051865
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2269809

  SDS
alfa aesar™
B2269809
10g Each for $57.57

AAB2269818

  SDS
alfa aesar™
B2269818
50g Each for $209.45

AAB2269830

  SDS
alfa aesar™
B2269830
250g Each for $845.30
Description & Specifications

Specifications

Formula Weight 222.18
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 10g
TSCA TSCA
Assay ≥98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Allyltriethylammonium bromide
CAS 29443-23-0
Melting Point 236° to 238°C
MDL Number MFCD00051865
Health Hazard 1 Warning
Molecular Formula CH2=CHCH2N(CH2CH3)3Br
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.