Please login to your online account to display your discounted pricing

Allyltrichlorosilane, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  313220050

 View more versions of this product

Catalog No. AC313220050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 175.52
Quantity 5g
Color Clear colorless to slightly yellow
TSCA TSCA
RTECS Number VV1530000
EINECS Number 203-485-9
CAS Min % 94.0
CAS Max % 100.0
Refractive Index 1.4430 to 1.4470 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
Contact with water liberates toxic gas.
Name Note 95%
Molecular Formula C3H5Cl3Si
CAS 107-37-9
Chemical Name or Material Allyltrichlorosilane
MDL Number MFCD00000486
Linear Formula HCCHCHSiCl3
Assay Percent Range 95%
Infrared Spectrum Authentic
Synonym 3-Trichlorosilylpropene
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Allyltrichlorosilane
Name Note 95%
CAS 107-37-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C3H5Cl3Si
Linear Formula HCCHCHSiCl3
Formula Weight 175.52
MDL Number MFCD00000486
Synonym 3-Trichlorosilylpropene
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
Contact with water liberates toxic gas.

SDS & Safety Documents