Learn More
Allyltributyltin, 97%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC265550250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Allyltributyltin | |
96.0 | |
1.0680g/mL | |
88.0°C to 92.0°C (0.2mmHg) | |
97% | |
C15H32Sn | |
MFCD00010346 | |
Glass bottle | |
15, 289 | |
allyl tributyl stannane, allyl tributyltin, allyltri-n-butyltin, allyltributylstannane, allyltributyltin, stannane, tributyl-2-propenyl, tri-n-butyl-2-propenylstannane, tributyl prop-2-en-1-yl stannane, tributyl-2-propenylstannane, unii-aor8od4103 | |
YLGRTLMDMVAFNI-UHFFFAOYSA-N | |
CCCC[Sn](CCCC)(CCCC)CC=C | |
331.13 | |
331.1 | |
97% |
24850-33-7 | |
100.0 | |
Colorless | |
>110°C | |
Authentic | |
[CH3(CH2)3]3SnCH2CH=CH2 | |
04, 1922 | |
09,8; 11,15; 12,21; 14,14; 16,7; 17,12 | |
1.4850 to 1.4870 | |
25g | |
1.068 | |
tributyl(prop-2-enyl)stannane | |
90628 | |
Liquid |
Chemical Identifiers
24850-33-7 | |
331.13 | |
YLGRTLMDMVAFNI-UHFFFAOYSA-N | |
90628 | |
CCCC[Sn](CCCC)(CCCC)CC=C |
C15H32Sn | |
MFCD00010346 | |
allyl tributyl stannane, allyl tributyltin, allyltri-n-butyltin, allyltributylstannane, allyltributyltin, stannane, tributyl-2-propenyl, tri-n-butyl-2-propenylstannane, tributyl prop-2-en-1-yl stannane, tributyl-2-propenylstannane, unii-aor8od4103 | |
tributyl(prop-2-enyl)stannane |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 246-494-3
RUO – Research Use Only