Learn More
Allylthiourea, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 148800250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Allylthiourea | |
109-57-9 | |
100.0 | |
White to Beige | |
Authentic | |
Glass bottle | |
H2C=CHCH2NHCSNH2 | |
25 g | |
15, 9515 | |
Solubility in water: 67g/L (20°C). Other solubilities: 1370g/L methanol (20°C)- insoluble in benzene,soluble in ether,solutions of borax,benzoates,and urethane,slightly soluble in 70% alcohol | |
C=CCNC(=S)N | |
116.18 | |
CHEBI:74079 | |
98% |
98% | |
97.5 | |
70.0°C to 78.0°C | |
1.5% max. (50°C, 2 hrs) | |
98% | |
C4H8N2S | |
MFCD00004940 | |
04, 211 | |
allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin | |
HTKFORQRBXIQHD-UHFFFAOYSA-N | |
prop-2-enylthiourea | |
1549517 | |
116.18 | |
Crystals, Powder, Granules or Chunks |
Chemical Identifiers
109-57-9 | |
HTKFORQRBXIQHD-UHFFFAOYSA-N | |
1549517 | |
prop-2-enylthiourea |
MFCD00004940 | |
allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin | |
CHEBI:74079 | |
C=CCNC(=S)N |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
EINECSNumber : 203-683-5
RTECSNumber : YR8050000
TSCA : TSCA
RUO – Research Use Only