Please login to your online account to display your discounted pricing

Alfa Aesar™ Allylpentafluorobenzene, 98%

Chemical Identifiers

Chemical Name or Material Allylpentafluorobenzene
CAS 1736-60-3
Molecular Formula C9H5F5
Formula Weight 208.13
MDL Number MFCD00000301
UN Number UN1993
Beilstein 1971209
Synonym 2,3,4,5,6-Pentafluoro(allylbenzene)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1783406

  SDS
alfa aesar™
A1783406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1783414

  SDS
alfa aesar™
A1783414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.13
Molecular Formula C9H5F5
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point 42°C (108°F)
Assay 98%
MDL Number MFCD00000301
CAS 1736-60-3
Boiling Point 148° to 149°C
Synonym 2,3,4,5,6-Pentafluoro(allylbenzene)
Quantity 5g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN1993
Chemical Name or Material Allylpentafluorobenzene
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Beilstein 1971209
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.