missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Allylpalladium(II) chloride dimer, Pd 56.0% min

Manufacturer:  Alfa Aesar™ 1000506

 View more versions of this product

Catalog No. AA1000506



Specifications

Specifications

Allylpalladium(II) chloride dimer
Yellow
5g
C6H10Cl2Pd2
4124623
Pd 56.0% min.
PENAXHPKEVTBLF-UHFFFAOYSA-L
chloropalladium(1+);prop-1-ene
61538
Powder
Air & Moisture Sensitive
120°C (decomposition)
12012-95-2
MFCD00044874
allyl palladium chloride dimer
Very soluble in dichloromethane and dichloroethane; Slightly soluble in toluene, insoluble in water
[CH2-]C=C.[CH2-]C=C.Cl[Pd+].Cl[Pd+]
365.886
365.85

Chemical Identifiers

12012-95-2
365.886
PENAXHPKEVTBLF-UHFFFAOYSA-L
61538
[CH2-]C=C.[CH2-]C=C.Cl[Pd+].Cl[Pd+]
C6H10Cl2Pd2
MFCD00044874
allyl palladium chloride dimer
chloropalladium(1+);prop-1-ene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RT3510000

storageNote1 : 2°C to 8°C

Documents