missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allyloxy-tert-butyldimethylsilane, 98%, Thermo Scientific Chemicals

$58.30 - $58.30

Chemical Identifiers

CAS 105875-75-0
Molecular Formula C9H20OSi
MDL Number MFCD00239355
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55077AA
View Documents Promotion Details
Thermo Scientific Chemicals
H55077AA
5 mL
Each for $58.30
 
Description

Description

Allyloxy-tert-butyldimethylsilane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyloxy-tert-butyldimethylsilane is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water (0.50 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

105875-75-0
C9H20OSi
MFCD00239355

Specifications

120°C (50mmHg)
5 mL
H2C=CHCH2OSi(CH3)2C(CH3)3
UN1993
Sparingly soluble in water (0.50g/L at 25°C).
98%
0.811
105875-75-0
C9H20OSi
MFCD00239355
33°C (91°F)
172.34
1.421
Allyloxy-tert-butyldimethylsilane
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only