missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyloxy-tert-butyldimethylsilane, 98%

Manufacturer:  Alfa Aesar™ H55077AA

 View more versions of this product

Catalog No. AAH55077AA



Description

Description

Applications
Allyloxy-tert-butyldimethylsilane is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water (0.50 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Allyloxy-tert-butyldimethylsilane
C9H20OSi
MFCD00239355
Sparingly soluble in water (0.50g/L at 25°C).
98%
1.421
5mL
105875-75-0
H2C=CHCH2OSi(CH3)2C(CH3)3
UN1993
172.34
0.811
120°C (50mmHg)
33°C (91°F)

Chemical Identifiers

105875-75-0
C9H20OSi
MFCD00239355
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents