missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Allylmalonic acid, 98+%

$106.22 - $1,259.57

Chemical Identifiers

CAS 2583-25-7
Molecular Formula CH2=CHCH2CH(CO2H)2
Formula Weight 144.13
MDL Number MFCD00014347
Beilstein 1766382
Chemical Name or Material Allylmalonic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1744306 SDS Alfa Aesar™
A1744306
5g Each for $106.22
AAA1744322 SDS Alfa Aesar™
A1744322
100g Each for $1,259.57
AAA1744314 SDS Alfa Aesar™
A1744314
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2583-25-7
144.13
1766382
CH2=CHCH2CH(CO2H)2
MFCD00014347
Allylmalonic acid

Specifications

2583-25-7
144.13
1766382
5g
≥98%
CH2=CHCH2CH(CO2H)2
MFCD00014347
103° to 105°C
Allylmalonic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning