missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allylmagnesium chloride, 1.7M solution in THF, AcroSeal™, ACROS Organics™

$75.98 - $75.98

Chemical Identifiers

CAS 2622-05-1,109-99-9,592-42-7
CAS Min % 15.76,80.04
CAS Max % 19.96,84.25
Molecular Formula C3H5ClMg
Linear Formula H2C=CHCH2MgCl
Formula Weight 100.83
MDL Number MFCD00000473
Chemical Name or Material Allylmagnesium chloride, 1.7M Solution in THF
Assay 1.5 to 1.9 M
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC209671000 SDS Thermo Fisher Scientific
AC209671000
AcroSeal™ Glass Bottle in Tin Can 100mL Each for $75.98
AC209678000 SDS Thermo Fisher Scientific
AC209678000
AcroSeal™ Glass Bottle 800mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2622-05-1,109-99-9,592-42-7
19.96,84.25
H2C=CHCH2MgCl
MFCD00000473
1.5 to 1.9 M
15.76,80.04
C3H5ClMg
100.83
Allylmagnesium chloride, 1.7M Solution in THF

Specifications

2622-05-1,109-99-9,592-42-7
19.96,84.25
C3H5ClMg
H2C=CHCH2MgCl
0.9600g/mL
AcroSeal™ Glass Bottle in Tin Can
100mL
1.5 to 1.9 M
15.76,80.04
Liquid
1.5 to 1.9M
100.83
MFCD00000473
Clear tan to brown, amber, dark gray, dark green or black
Allylmagnesium chloride, 1.7M Solution in THF

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. Suspected of causing cancer. Highly flammable liquid and vapor. Reacts violently with water. May form explosive peroxides. In contact with water releases flammable gases which may ignite spontaneously. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger