Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyldiphenylphosphine, 96%

$88.76 - $88.76

Chemical Identifiers

Chemical Name or Material Allyldiphenylphosphine
CAS 2741-38-0
Linear Formula (CH2=CHCH2)P(C6H5)2
Formula Weight 226.25
MDL Number MFCD00014954
UN Number UN3278
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3030309

  SDS
alfa aesar™
3030309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3030304

  SDS
alfa aesar™
3030304
2g Each for $88.76
Description & Specifications

Specifications

Formula Weight 226.25
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Allyldiphenylphosphine
Physical Form Liquid
Assay 96%
Linear Formula (CH2=CHCH2)P(C6H5)2
Refractive Index 1.6190
MDL Number MFCD00014954
CAS 2741-38-0
Density 1.049g/mL
UN Number UN3278
Health Hazard 1 Danger
Color Colorless
Boiling Point 194° to 200°C (15mmHg)
Quantity 10g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point >110° (230°F)
Sensitivity Air Sensitive
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.