Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyldichloromethylsilane, 95%

Chemical Identifiers

Chemical Name or Material Allyldichloromethylsilane
CAS 1873-92-3
Molecular Formula C4H8Cl2Si
Formula Weight 155.1
MDL Number MFCD00045141
UN Number UN2985
Beilstein 1740877
Synonym Allylmethyldichlorosilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0304106

 
alfa aesar™
L0304106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0304114

 
alfa aesar™
L0304114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 155.1
Chemical Name or Material Allyldichloromethylsilane
Molecular Formula C4H8Cl2Si
CAS 1873-92-3
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay 95%
MDL Number MFCD00045141
UN Number UN2985
Beilstein 1740877
Synonym Allylmethyldichlorosilane
Quantity 5g
Flash Point 13°C (55°F)
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.
TSCA TSCA
Odor Acrid
Boiling Point 119° to 120°C

Applications
Allyldichloromethylsilane is used as Chemical Additives and pharmaceutical intermediates.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Incompatible with acids, Water, Bases, Ketones, Aldehydes, Alcohols, Peroxides, Amines, Ammonia, Oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.