Please login to your online account to display your discounted pricing

Alfa Aesar™ Allylcyclohexane, 96%

Chemical Identifiers

Chemical Name or Material Allylcyclohexane
CAS 2114-42-3
Molecular Formula C9H16
Formula Weight 124.23
MDL Number MFCD00013769
UN Number UN3295
Beilstein 2037346
Synonym 3-Cyclohexyl-1-propene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0094606

  SDS
alfa aesar™
L0094606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0094614

  SDS
alfa aesar™
L0094614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 124.23
Chemical Name or Material Allylcyclohexane
Assay 96%
CAS 2114-42-3
Health Hazard 2 GHS H Statement
H304-H318-H226
May be fatal if swallowed and enters airways.
Causes serious eye damage.
Flammable liquid and vapor.
Synonym 3-Cyclohexyl-1-propene
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00013769
Quantity 5g
UN Number UN3295
Flash Point 31°C (87°F)
Boiling Point 153° to 154°C
Molecular Formula C9H16
Beilstein 2037346
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.