Learn More
Allylchlorodimethylsilane, 94%
CAS: 4028-23-3 | C5H11ClSi | 134.678 g/mol
Supplier: Thermo Scientific Chemicals B2223709
Description
Allylchlorodimethylsilane is used in the synthesis of cyclo-1,1?,4,4?-bis(1,1,3,3-tetramethyl-1,3-disiloxanediyl)dibenzene, silylating agent for steroids for analysis by GC-MS. Also used in the preparation of phenylenebis(silanediyl triflates), useful synthons for organosilicon polymers, O-silylation of phenyl sulfones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAllylchlorodimethylsilane is used in the synthesis of cyclo-1,1′,4,4′-bis(1,1,3,3-tetramethyl-1,3-disiloxanediyl)dibenzene, silylating agent for steroids for analysis by GC-MS. Also used in the preparation of phenylenebis(silanediyl triflates), useful synthons for organosilicon polymers, O-silylation of phenyl sulfones.
Solubility
Soluble in most organic solvents; reacts with alcohols, amines, and water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, strong bases, water/ moisture.
Specifications
| 110°C to 112°C | |
| 4028-23-3 | |
| C5H11ClSi | |
| UN2985 | |
| 5°C (41°F) | |
| Soluble in most organic solvents; reacts with alcohols,amines,and water. | |
| C[Si](C)(CC=C)Cl | |
| 134.678 | |
| 134.68 | |
| 1.429 | |
| 0.896 |
| Allylchlorodimethylsilane | |
| 10 g | |
| MFCD00000506 | |
| 1737978 | |
| allyldimethylchlorosilane, allylchlorodimethylsilane, silane, chlorodimethyl-2-propenyl, allyl chloro dimethylsilane, allyl-chloro-dimethyl silane, silane, chlorodimethyl-2-propen-1-yl, admcs, allyl chlorodimethylsilane, allylchlorodimethyl silane, acmc-1afr7 | |
| KMVZWUQHMJAWSY-UHFFFAOYSA-N | |
| chloro-dimethyl-prop-2-enylsilane | |
| 77646 | |
| 97% | |
| Moisture sensitive |
Chemical Identifiers
| 4028-23-3 | |
| 134.678 | |
| KMVZWUQHMJAWSY-UHFFFAOYSA-N | |
| 77646 | |
| C[Si](C)(CC=C)Cl |
| C5H11ClSi | |
| MFCD00000506 | |
| allyldimethylchlorosilane, allylchlorodimethylsilane, silane, chlorodimethyl-2-propenyl, allyl chloro dimethylsilane, allyl-chloro-dimethyl silane, silane, chlorodimethyl-2-propen-1-yl, admcs, allyl chlorodimethylsilane, allylchlorodimethyl silane, acmc-1afr7 | |
| chloro-dimethyl-prop-2-enylsilane |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 223-706-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only