Please login to your online account to display your discounted pricing

Allylamine, 98+%, extra pure, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  220732500

 View more versions of this product

Catalog No. AC220732500

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Extra Pure
Physical Form Crystalline or Granular Powder
Assay ≥97.5% (GC)
Formula Weight 57.09
Quantity 250mL
Color White or almost white
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECS Number 203-463-9
TSCA TSCA
RTECS Number BA5425000
CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4200 to 1.4230 (20°C, 589nm)
Water 0.5% max.
Health Hazard 2 GHS P Statement
Highly flammable liquid and vapor.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Chemical Name or Material Allylamine
Name Note 98+%
Assay Percent Range 98%
Molecular Formula C3H7N
Linear Formula H2C=CHCH2NH2
MDL Number MFCD00008199
CAS 107-11-9
Synonym 3-Amino-1-propene
Health Hazard 1 Danger

Specifications

Chemical Name or Material Allylamine
Name Note 98+%
CAS 107-11-9
CAS Min % 98.0
CAS Max % 100.0
Purity Grade Extra Pure
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C3H7N
Linear Formula H2C=CHCH2NH2
Formula Weight 57.09
MDL Number MFCD00008199
Synonym 3-Amino-1-propene
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Highly flammable liquid and vapor.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.

SDS & Safety Documents