Please login to your online account to display your discounted pricing

Allylamine, 98+%, extra pure, ACROS Organics™


Quantity 50mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  220730500

 View more versions of this product

Catalog No. AC220730500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (GC)
Purity Grade Extra Pure
Physical Form Crystalline or Granular Powder
Formula Weight 57.09
Quantity 50mL
Color White or almost white
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECS Number 203-463-9
TSCA TSCA
RTECS Number BA5425000
CAS Min % 98.0
CAS Max % 100.0
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Highly flammable liquid and vapor.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Chemical Name or Material Allylamine
Name Note 98+%
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C3H7N
Linear Formula H2C=CHCH2NH2
Refractive Index 1.4200 to 1.4230 (20°C, 589nm)
MDL Number MFCD00008199
CAS 107-11-9
Infrared Spectrum Authentic
Synonym 3-Amino-1-propene
Water 0.5% max.

Specifications

Chemical Name or Material Allylamine
Name Note 98+%
CAS 107-11-9
CAS Min % 98.0
CAS Max % 100.0
Purity Grade Extra Pure
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C3H7N
Linear Formula H2C=CHCH2NH2
Formula Weight 57.09
MDL Number MFCD00008199
Synonym 3-Amino-1-propene
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Highly flammable liquid and vapor.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.

SDS & Safety Documents