Please login to your online account to display your discounted pricing

Allylamine, 98+%, extra pure, ACROS Organics™

$72.65 - $72.65

Chemical Identifiers

Chemical Name or Material Allylamine
Name Note 98+%
CAS 107-11-9
CAS Min % 98.0
CAS Max % 100.0
Purity Grade Extra Pure
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C3H7N
Linear Formula H2C=CHCH2NH2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC220730500

  SDS
thermo fisher scientific
220730500
Glass bottle 50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC220732500

  SDS
thermo fisher scientific
220732500
Glass bottle 250mL Each for $72.65
Description & Specifications

Specifications

Assay ≥97.5% (GC)
Formula Weight 57.09
Purity Grade Extra Pure
Physical Form Crystalline or Granular Powder
Quantity 50mL
Color White or almost white
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
TSCA TSCA
RTECS Number BA5425000
EINECS Number 203-463-9
CAS Min % 98.0
CAS Max % 100.0
Health Hazard 2 GHS P Statement
Highly flammable liquid and vapor.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Health Hazard 1 Danger
Chemical Name or Material Allylamine
Molecular Formula C3H7N
Linear Formula H2C=CHCH2NH2
Assay Percent Range 98%
CAS 107-11-9
Name Note 98+%
MDL Number MFCD00008199
Packaging Glass bottle
Refractive Index 1.4200 to 1.4230 (20°C, 589nm)
Water 0.5% max.
Infrared Spectrum Authentic
Synonym 3-Amino-1-propene
Safety and Handling

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Highly flammable liquid and vapor.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.