Please login to your online account to display your discounted pricing

Alfa Aesar™ Allylamine, 98+%

$62.39 - $62.39

Chemical Identifiers

Chemical Name or Material Allylamine
CAS 107-11-9
Molecular Formula C3H7N
Linear Formula CH2=CHCH2NH2
Formula Weight 57.1
MDL Number MFCD00008199
UN Number UN2334
Beilstein 635703
Merck Index 14,287
Synonym 3-Amino-1-propene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL07162AE

  SDS
alfa aesar™
L07162AE
100mL Each for $62.39

AAL07162AC

  SDS
alfa aesar™
L07162AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL07162AP

  SDS
alfa aesar™
L07162AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 57.1
Odor Ammonia-like
Assay ≥98%
RTECS Number BA5425000
TSCA TSCA
Merck Index 14,287
Melting Point −88°C
Quantity 100mL
Boiling Point 53° to 54°C
CAS 107-11-9
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00008199
Health Hazard 1 Danger
Molecular Formula C3H7N
Health Hazard 2 GHS H Statement
H225-H301-H311-H331
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Flash Point −28°C (−18°F)
UN Number UN2334
Beilstein 635703
Linear Formula CH2=CHCH2NH2
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALLYLAMINE
Synonym 3-Amino-1-propene
Chemical Name or Material Allylamine

Applications
Allylamine is used as monomer in deposition of the allylamine plasma polymer. It was used in generation of a chemical gradient via plasma co-polymerization of octadiene and allylamine polymer on nanoporous alumina. It was used to investigate the surface reactivity of substrates coated with thin layers of plasma polymerized allylamine films. It was used in a direct monocarboxymethylation reaction.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store away from air and oxidizing agents. Store under dry inert gas. Incompatible with air and oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H301-H311-H331
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.