missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allyl Trifluoroacetate, TCI America™

Manufacturer:  TCI America T176525G

 View more versions of this product

Catalog No. T176525G



Specifications

Specifications

Allyl Trifluoroacetate
Colorless
383-67-5
MFCD00013567
allyl trifluoroacetate, allyl trifluoracetate, trifluoroacetic acid allyl ester, prop-2-en-1-yl 2,2,2-trifluoroacetate, allyltrifluoroacetate, pubchem12548, acmc-209izu, allyltrifluoroacetate, allyl alcohol, trifluoroacetate, acetic acid, trifluoro-, allyl ester
C=CCOC(=O)C(F)(F)F
154.088
154.09
≥95.0% (GC)
66°C
25g
C5H5F3O2
2924
XIVPVSIDXBTZLM-UHFFFAOYSA-N
prop-2-enyl 2,2,2-trifluoroacetate
67845
Clear Liquid at 20°C

Chemical Identifiers

383-67-5
154.088
XIVPVSIDXBTZLM-UHFFFAOYSA-N
67845
C=CCOC(=O)C(F)(F)F
C5H5F3O2
MFCD00013567
allyl trifluoroacetate, allyl trifluoracetate, trifluoroacetic acid allyl ester, prop-2-en-1-yl 2,2,2-trifluoroacetate, allyltrifluoroacetate, pubchem12548, acmc-209izu, allyltrifluoroacetate, allyl alcohol, trifluoroacetate, acetic acid, trifluoro-, allyl ester
prop-2-enyl 2,2,2-trifluoroacetate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents