missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allyl Sulfide, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC103021000

 View more versions of this product

Catalog No. AC103021000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Allyl sulfide
96.0
Authentic
1.4879 to 1.4899 (20°C, 589nm)
≥96.0%
C6H10S
0.887g/mL
MFCD00008658
Clear colorless
100mL
592-88-1
100.0
Glass bottle
Liquid
97%
(H2C=CHCH2)2S
114.2
01, 440
15, 288

Chemical Identifiers

592-88-1
100.0
C6H10S
114.2
Allyl sulfide
96.0
97%
(H2C=CHCH2)2S
MFCD00008658
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning