Please login to your online account to display your discounted pricing

Allyl Sulfide, 97%, ACROS Organics™

$34.18 - $34.18

Chemical Identifiers

Chemical Name or Material Allyl sulfide
CAS 592-88-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H10S
Linear Formula (H2C=CHCH2)2S
Formula Weight 114.2
MDL Number MFCD00008658
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC103021000

  SDS
thermo fisher scientific
AC103021000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC103020250

  SDS
thermo fisher scientific
103020250
Glass bottle 25mL Each for $34.18
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 114.2
Quantity 100mL
Color Clear colorless
Density 0.887g/mL
Beilstein 01, 440
Merck Index 15, 288
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula (H2C=CHCH2)2S
Refractive Index 1.4879 to 1.4899 (20°C, 589nm)
Molecular Formula C6H10S
MDL Number MFCD00008658
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Chemical Name or Material Allyl sulfide
CAS 592-88-1
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.