Please login to your online account to display your discounted pricing

Allyl Sulfide, 97%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  103020250

 View more versions of this product

Catalog No. AC103020250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 114.2
Physical Form Liquid
Quantity 25mL
Color Clear colorless
Beilstein 01, 440
Merck Index 15, 288
CAS Max % 100.0
CAS Min % 96.0
Linear Formula (H2C=CHCH2)2S
Refractive Index 1.4879 to 1.4899 (20°C, 589nm)
Molecular Formula C6H10S
MDL Number MFCD00008658
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Allyl sulfide
CAS 592-88-1
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Allyl sulfide
CAS 592-88-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H10S
Linear Formula (H2C=CHCH2)2S
Formula Weight 114.2
MDL Number MFCD00008658
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

SDS & Safety Documents