missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Allyl phenyl sulfone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1040106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Allyl phenyl sulfone | |
1.189 | |
>110°C (230°F) | |
MFCD00014739 | |
1863561 | |
allyl phenyl sulfone, benzene, 2-propenylsulfonyl, allylsulfonyl benzene, phenyl allyl sulfone, sulfone, allyl phenyl, allylsulphonyl benzene, allylphenyl sulfone, 3-phenylsulfonyl-1-propene, benzene, 2-propen-1-ylsulfonyl | |
C=CCS(=O)(=O)C1=CC=CC=C1 | |
182.237 | |
182.24 |
16212-05-8 | |
111°C to 113°C (0.5 mmHg) | |
C9H10O2S | |
5 g | |
Light sensitive | |
KYPIULIVYSQNNT-UHFFFAOYSA-N | |
prop-2-enylsulfonylbenzene | |
85330 | |
98% |
Chemical Identifiers
16212-05-8 | |
182.237 | |
KYPIULIVYSQNNT-UHFFFAOYSA-N | |
85330 | |
C=CCS(=O)(=O)C1=CC=CC=C1 |
C9H10O2S | |
MFCD00014739 | |
allyl phenyl sulfone, benzene, 2-propenylsulfonyl, allylsulfonyl benzene, phenyl allyl sulfone, sulfone, allyl phenyl, allylsulphonyl benzene, allylphenyl sulfone, 3-phenylsulfonyl-1-propene, benzene, 2-propen-1-ylsulfonyl | |
prop-2-enylsulfonylbenzene |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 240-338-8
RTECSNumber : WR2400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only