missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl methyl ether, 94%

Manufacturer:  Alfa Aesar™ 1865218

 View more versions of this product

Catalog No. AA1865218



Specifications

Specifications

Allyl methyl ether
−23° (−94°F)
1.3803
50g
C4H8O
UN3271
FASUFOTUSHAIHG-UHFFFAOYSA-N
3-methoxyprop-1-ene
69392
Liquid
0.77g/mL
Ether-like
42°C to 43°C
627-40-7
MFCD00008649
allyl methyl ether, 1-propene, 3-methoxy, 3-methoxy-1-propene, ether, allyl methyl, 3-methoxypropene, methyl allyl ether, allylmethylether, 4-oxapent-1-ene, 1-methoxy-2-propene, methylallyl ether
COCC=C
72.107
72.1
94%

Chemical Identifiers

627-40-7
72.107
FASUFOTUSHAIHG-UHFFFAOYSA-N
69392
COCC=C
C4H8O
MFCD00008649
allyl methyl ether, 1-propene, 3-methoxy, 3-methoxy-1-propene, ether, allyl methyl, 3-methoxypropene, methyl allyl ether, allylmethylether, 4-oxapent-1-ene, 1-methoxy-2-propene, methylallyl ether
3-methoxyprop-1-ene
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

TSCA : TSCA

SDS Documents