Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl methacrylate, 97%, stab. with 100 ppm 4-methoxyphenol

Chemical Identifiers

Chemical Name or Material Allyl methacrylate
CAS 96-05-9
Name Note stabilized with 100ppm 4-methoxyphenol
Molecular Formula C7H10O2
Formula Weight 126.16
MDL Number MFCD00008592
UN Number UN2929
Beilstein 1747406
Synonym Methacrylic acid allyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1299318

 
alfa aesar™
L1299318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1299330

 
alfa aesar™
L1299330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.16
Chemical Name or Material Allyl methacrylate
Assay 97%
Molecular Formula C7H10O2
MDL Number MFCD00008592
CAS 96-05-9
TSCA TSCA
Name Note stabilized with 100ppm 4-methoxyphenol
UN Number UN2929
Beilstein 1747406
Melting Point −65°C
Synonym Methacrylic acid allyl ester
Boiling Point 59° to 60°C (43mmHg)
Quantity 50g
Flash Point 33°C (91°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H330-H226-H302
Toxic in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
RTECS Number UD3483000
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H311-H330-H226-H302
Toxic in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.