missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allyl mercaptan, tech., 70%, remainder mainly sulfide, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 153710250

 View more versions of this product

Catalog No. AC153710250



Specifications

Specifications

Allyl mercaptan
18°C
Glass bottle
0.93
Colorless to Yellow
870-23-5
H2C=CHCH2SH
01, 440
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
C=CCS
74.13
74.13
70%
0.9300g/mL
Authentic
1.4745 to 1.4785
67°C to 68°C
25g
C3H6S
MFCD00004894
2-propene-1-thiol
ULIKDJVNUXNQHS-UHFFFAOYSA-N
prop-2-ene-1-thiol
13367
Liquid
Technical

Chemical Identifiers

870-23-5
74.13
ULIKDJVNUXNQHS-UHFFFAOYSA-N
13367
C=CCS
C3H6S
MFCD00004894
2-propene-1-thiol
prop-2-ene-1-thiol
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Highly flammable liquid and vapor

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Danger

EINECSNumber : 212-792-7

TSCA : TSCA

SDS Documents