Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl mercaptan, tech. 70%

$90.96 - $90.96

Chemical Identifiers

Chemical Name or Material Allyl mercaptan
CAS 870-23-5
Purity Grade Technical Grade
Molecular Formula C3H6S
Linear Formula CH2=CHCH2SH
Formula Weight 74.15
MDL Number MFCD00004894
UN Number UN1228
Beilstein 1697523
Synonym 2-Propene-1-thiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1361614

 
alfa aesar™
A1361614
25g Each for $90.96

AAA1361622

 
alfa aesar™
A1361622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 74.15
Chemical Name or Material Allyl mercaptan
Odor Stench
TSCA TSCA
Purity Grade Technical Grade
Assay 70%
Molecular Formula C3H6S
Linear Formula CH2=CHCH2SH
MDL Number MFCD00004894
CAS 870-23-5
UN Number UN1228
Beilstein 1697523
Synonym 2-Propene-1-thiol
Boiling Point 67° to 68°C
Quantity 25g
Flash Point 18°C (64°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H331-H319-H335
Highly flammable liquid and vapour.
Toxic if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P260-P280f-P305+P351+P338-P403+P233
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place.
Keep container tightly closed.
Recommended Storage −30° to −10°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, TOXIC, N.O.S.

Applications
Allyl mercaptan is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. It is sensitive to air. Incompatible with oxidizing agents. Store under dry inert gas.

Safety and Handling

Danger

GHS P Statement
P210-P260-P280f-P305+P351+P338-P403+P233
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place.
Keep container tightly closed.

GHS H Statement
H225-H331-H319-H335
Highly flammable liquid and vapour.
Toxic if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C