missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl isocyanate, 98%

Manufacturer:  Alfa Aesar™ L1871006

 View more versions of this product

Catalog No. AAL1871006



Description

Description

Applications
It finds its application in the synthesis, characterization of biodegradable dextran-allyl isocyanate-ethylamine/polyethylene glycol-diacrylate hydrogels and their in vitro release of albumin.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from alcohols, amines, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges. Store under inert gas.
Specifications

Specifications

Allyl isocyanate
Pungent
5g
C4H5NO
UN2478
1-propene, 3-isocyanato
C=CCN=C=O
83.09
83.09
6°C (43°F)
87°C to 89°C
1476-23-9
MFCD00002044
506106
HXBPYFMVGFDZFT-UHFFFAOYSA-N
3-isocyanatoprop-1-ene
15123
98%

Chemical Identifiers

1476-23-9
83.09
HXBPYFMVGFDZFT-UHFFFAOYSA-N
15123
C=CCN=C=O
C4H5NO
MFCD00002044
1-propene, 3-isocyanato
3-isocyanatoprop-1-ene
Safety and Handling

Safety and Handling

GHS H Statement
H225-H300-H330-H334-H314-H318
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOCYANATES, FLAMMABLE, TOXIC, N.O.S.

RTECSNumber : NQ8175000

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents