Please login to your online account to display your discounted pricing

Allyl Iodide, Stabilized 98%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC218545000

 View more versions of this product

Catalog No. AC218545000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5% (GC)
Formula Weight 167.98
Quantity 500mL
Color Clear yellow to orange, red or red-brown
Density 1.83g/mL
Chemical Name or Material Allyl iodide
Assay Percent Range 98%
Molecular Formula C3H5I
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula H2C=CHCH2I
Refractive Index 1.5525 to 1.5555 (20°C, 589nm)
CAS 556-56-9
Stabilizer Copper

Specifications

Chemical Name or Material Allyl iodide
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C3H5I
Linear Formula H2C=CHCH2I
Formula Weight 167.98