Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl heptanoate, 97%

Chemical Identifiers

Chemical Name or Material Allyl heptanoate
CAS 142-19-8
Molecular Formula C10H18O2
Linear Formula CH3(CH2)5CO2CH2CH=CH2
Formula Weight 170.25
MDL Number MFCD00038341
UN Number UN2810
Beilstein 8544440
Synonym Heptanoic acid allyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1067136

 
alfa aesar™
L1067136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1067122

 
alfa aesar™
L1067122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.25
Chemical Name or Material Allyl heptanoate
Molecular Formula C10H18O2
MDL Number MFCD00038341
Synonym Heptanoic acid allyl ester
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 97%
Linear Formula CH3(CH2)5CO2CH2CH=CH2
CAS 142-19-8
UN Number UN2810
Beilstein 8544440
Quantity 500g
Health Hazard 2 GHS H Statement
H302-H312-H315-H319-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECS Number MJ1750000
Health Hazard 1 Warning
Boiling Point 210° to 212°C
Flash Point 74°C (165°F)
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H315-H319-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.