missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl heptanoate, 97%

Manufacturer:  Alfa Aesar™ L1067122

 View more versions of this product

Catalog No. AAL1067122



Specifications

Specifications

Allyl heptanoate
210°C to 212°C
142-19-8
CH3(CH2)5CO2CH2CH=CH2
UN2810
allyl heptanoate, allyl heptylate, 2-propenyl heptanoate, allyl enanthate, allyl heptoate, heptanoic acid, 2-propenyl ester, prop-2-en-1-yl heptanoate, heptanoic acid, allyl ester, allylester kyseliny enanthove, heptanoic acid, 2-propen-1-yl ester
CCCCCCC(=O)OCC=C
170.252
170.25
74°C (165°F)
100g
C10H18O2
MFCD00038341
8544440
SJWKGDGUQTWDRV-UHFFFAOYSA-N
prop-2-enyl heptanoate
8878
97%

Chemical Identifiers

142-19-8
170.252
SJWKGDGUQTWDRV-UHFFFAOYSA-N
8878
CCCCCCC(=O)OCC=C
C10H18O2
MFCD00038341
allyl heptanoate, allyl heptylate, 2-propenyl heptanoate, allyl enanthate, allyl heptoate, heptanoic acid, 2-propenyl ester, prop-2-en-1-yl heptanoate, heptanoic acid, allyl ester, allylester kyseliny enanthove, heptanoic acid, 2-propen-1-yl ester
prop-2-enyl heptanoate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : MJ1750000

TSCA : TSCA

Documents