Learn More
Allyl Glycidyl Ether, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 102941000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Allyl glycidyl ether | |
106-92-3 | |
100.0 | |
154.0°C | |
Authentic | |
Glass bottle | |
1.4332 to 1.4352 | |
100g | |
allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi | |
LSWYGACWGAICNM-UHFFFAOYSA-N | |
2-(prop-2-enoxymethyl)oxirane | |
7838 | |
99+% |
99+% | |
99.0 | |
0.9620g/mL | |
48°C | |
99% min. (GC) | |
C6H10O2 | |
MFCD00005143 | |
0.962 | |
Solubility in water: 50g/L (20°C). Other solubilities: soluble in ethanol,acetone,diethyl ether and,benzene | |
C=CCOCC1CO1 | |
114.14 | |
114.14 |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Suspected of causing cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility.
Toxic if inhaled.
Harmful if swallowed.
May cause respiratory irri
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
EINECSNumber : 203-442-4
RTECSNumber : RR0875000
TSCA : TSCA
RUO â Research Use Only