missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allyl glycidyl ether, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1163422

 View more versions of this product

Catalog No. AAL1163422



Description

Description

Allyl glycidyl ether is useful in hydrosilylation, including the H2PtCl6·xH2O (cat. no. 206083) catalyzed hydrosilylation of siloxanes to epoxysiloxanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl glycidyl ether is useful in hydrosilylation, including the H2PtCl6·xH2O (cat. no. 206083) catalyzed hydrosilylation of siloxanes to epoxysiloxanes.

Solubility
Soluble in water (140 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Allyl glycidyl ether
-100°C
50°C to 51°C (15 mmHg)
Ether-like
1.435
100 g
105871
Soluble in water (140g/L at 20°C).
C=CCOCC1CO1
114.144
114.15
106-92-3
0.967
47°C (134°F)
C6H10O2
MFCD00005143
UN2219
allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
LSWYGACWGAICNM-UHFFFAOYSA-N
2-(prop-2-enoxymethyl)oxirane
7838
97%

Chemical Identifiers

106-92-3
114.144
LSWYGACWGAICNM-UHFFFAOYSA-N
7838
C=CCOCC1CO1
C6H10O2
MFCD00005143
allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
2-(prop-2-enoxymethyl)oxirane
Safety and Handling

Safety and Handling

GHS H Statement
H318-H226-H341-H351-H361-H302-H332-H317-H335-H315
Causes serious eye damage.
Flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280g-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P311-P330-P333+P313-P363-P37

H226-H302-H315-H317-H318-H331-H335-H341-H351-H361f-H373

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALLYL GLYCIDYL ETHER

EINECSNumber : 203-442-4

RTECSNumber : RR0875000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only