missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl cyanide, 97%

Manufacturer:  Alfa Aesar™ 4477522

 View more versions of this product

Catalog No. AA4477522



Specifications

Specifications

Allyl cyanide
24°C (75°F)
1.4050
Brown to Colorless
100g
C4H5N
MFCD00001962
605352
3-butenenitrile, allyl cyanide, 3-butenylnitrile, 3-butenonitrile, allylnitrile, vinylacetonitrile, 3-cyanopropene, 3-cyano-1-propene, 1-butene-4-nitrile, propene-3-nitrile
C=CCC#N
67.091
67.09
97%
0.834g/mL
Unpleasant
118°C to 120°C
-87°C
109-75-1
CH2=CHCH2CN
UN1992
14,290
SJNALLRHIVGIBI-UHFFFAOYSA-N
but-3-enenitrile
8009
Liquid

Chemical Identifiers

109-75-1
67.091
SJNALLRHIVGIBI-UHFFFAOYSA-N
8009
C=CCC#N
C4H5N
MFCD00001962
3-butenenitrile, allyl cyanide, 3-butenylnitrile, 3-butenonitrile, allylnitrile, vinylacetonitrile, 3-cyanopropene, 3-cyano-1-propene, 1-butene-4-nitrile, propene-3-nitrile
but-3-enenitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H226-H361-H312-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : EM8050000

TSCA : TSCA

SDS Documents