Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl crotonate, 98%

Chemical Identifiers

Chemical Name or Material Allyl crotonate
CAS 20474-93-5
Molecular Formula CH3CH=CHCO2CH2CH=CH2
Formula Weight 126.16g/mol
MDL Number MFCD00026990
UN Number UN3272
Beilstein 1751826
Synonym Crotonic acid allyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0341406

 
alfa aesar™
L03414-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 63° to 65°C (20mmHg)
Quantity 5g
Flash Point 48°C (118°F)
Chemical Name or Material Allyl crotonate
CAS 20474-93-5
Assay 98%
Molecular Formula CH3CH=CHCO2CH2CH=CH2
Formula Weight 126.16g/mol
MDL Number MFCD00026990
UN Number UN3272
Beilstein 1751826
Synonym Crotonic acid allyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.