Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl(chloro)(di-tert-butylneopentylphosphine)palladium(II), may cont. up to ca 5% toluene

Chemical Identifiers

Chemical Name or Material Allyl(chloro)(di-tert-butylneopentylphosphine)palladium(II)
CAS 1246775-56-3
Name Note May contain up to ≈5% toluene
Formula Weight 399.29
MDL Number MFCD25372544
Synonym PdClAllyl(dtbnp); Allyl[di-(tert-butyl)neopentylphosphine]palladium(II) chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4652903

  SDS
alfa aesar™
4652903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Allyl(chloro)(di-tert-butylneopentylphosphine)palladium(II)
Name Note May contain up to ≈5% toluene
CAS 1246775-56-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
Physical Form Crystalline Solid
Formula Weight 399.29
MDL Number MFCD25372544
Synonym PdClAllyl(dtbnp); Allyl[di-(tert-butyl)neopentylphosphine]palladium(II) chloride
Color Yellow
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.