Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl(chloro)[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II)

$397.78 - $397.78

Chemical Identifiers

Chemical Name or Material Allyl (chloro)[di-tert-butyl (4-dimethylaminophenyl)phosphine]palladium (II)
CAS 1235509-04-2
Molecular Formula C19H33ClNPPd
Formula Weight 448.32
MDL Number MFCD25372543
Synonym PdClAllyl(Amphos)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4633203

  SDS
alfa aesar™
4633203
1g Each for $397.78
Description & Specifications

Specifications

Chemical Name or Material Allyl (chloro)[di-tert-butyl (4-dimethylaminophenyl)phosphine]palladium (II)
CAS 1235509-04-2
Physical Form Solid
Molecular Formula C19H33ClNPPd
Formula Weight 448.32
MDL Number MFCD25372543
Synonym PdClAllyl(Amphos)
Color Yellow
Quantity 1g