Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl(chloro)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)

Chemical Identifiers

Chemical Name or Material Allyl(chloro)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)
CAS 1248656-98-5
Formula Weight 893.67
MDL Number MFCD25372546
Synonym PdClAllyl(Qphos)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4674703

  SDS
alfa aesar™
4674703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 893.67
Color Pink
Physical Form Solid
CAS 1248656-98-5
MDL Number MFCD25372546
Synonym PdClAllyl(Qphos)
Quantity 1g
Chemical Name or Material Allyl(chloro)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)