missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allyl chloride 98%, stabilized, ACROS Organics™

Manufacturer:  Acros Organics 102910010

 View more versions of this product

Catalog No. AC102910010



Description

Description

Alkylation reagent
Specifications

Specifications

Allyl chloride
−29°C
Glass bottle
0.939
0.34 mPa.s (20°C)
44.0°C to 46.0°C
-136.0°C
107-05-1,75-56-9
100.0
C3H5Cl
MFCD00000984
13,198
1-chloro-2-propene
OSDWBNJEKMUWAV-UHFFFAOYSA-N
3-chloroprop-1-ene
7850
76.53
98%
0.9390g/mL
Authentic
1.4140 to 1.4160
0.05 to 0.09% propylene oxide
0.02% max. (K.F.)
Colorless to Yellow
1L
97.5
98%
H2C=CHCH2Cl
01,198
15,28
Solubility in water: 3.6g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol>50g/100mL,soluble in ch2cl2,acetone >50g/100 mL,soluble in n,n-dimethylformamide >50g/100 mL,miscible with chloroform and ether
C=CCCl
76.53
CHEBI:82379
Liquid

Chemical Identifiers

107-05-1
76.53
OSDWBNJEKMUWAV-UHFFFAOYSA-N
7850
3-chloroprop-1-ene
C3H5Cl
MFCD00000984
1-chloro-2-propene
CHEBI:82379
C=CCCl
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation. Suspected of causing cancer. Suspected of causing genetic defects. Harmful in contact with skin. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Causes skin irritation. May cause respiratory irritation. Harmful if inhaled. Very toxic to aquatic life. Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Use personal protective equipment as required. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. Take precautionary measures against static discharge. Store in a well-ventilated place. Keep cool.

Danger

SDS Documents