Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl acrylate, tech. 90%, stab. with ca 100ppm 4-methoxyphenol

Chemical Identifiers

Chemical Name or Material Allyl acrylate
CAS 999-55-3
Name Note stabilized with ≈100ppm 4-methoxyphenol
Purity Grade Technical Grade
Molecular Formula CH2=CHCOOCH2CH=CH2
Formula Weight 112.13
MDL Number MFCD00014949
UN Number UN2929
Beilstein 1743576
Synonym Acrylic acid allyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1013806

  SDS
alfa aesar™
L1013806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1013814

  SDS
alfa aesar™
L1013814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.13
MDL Number MFCD00014949
Molecular Formula CH2=CHCOOCH2CH=CH2
Health Hazard 2 GHS H Statement
H310-H330-H226-H302-H315-H319-H335
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 90%
Beilstein 1743576
Odor Pungent
Name Note stabilized with ≈100ppm 4-methoxyphenol
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Acrylic acid allyl ester
Quantity 5g
RTECS Number UD3675000
TSCA TSCA
Health Hazard 1 Danger
Boiling Point 46° to 47°C (40mmHg)
UN Number UN2929
Purity Grade Technical Grade
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
Flash Point 32°C (89°F)
Chemical Name or Material Allyl acrylate
CAS 999-55-3
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H310-H330-H226-H302-H315-H319-H335
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.