missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl acrylate, tech. 90%, stab. with ca 100ppm 4-methoxyphenol

Manufacturer:  Alfa Aesar™ L1013814

 View more versions of this product

Catalog No. AAL1013814



Specifications

Specifications

Allyl acrylate
Pungent
25g
999-55-3
MFCD00014949
1743576
QTECDUFMBMSHKR-UHFFFAOYSA-N
prop-2-enyl prop-2-enoate
13835
90%
32°C (89°F)
46°C to 47°C (40mmHg)
stabilized with ≈100ppm 4-methoxyphenol
C6H8O2
UN2929
allyl acrylate, acrylic acid, allyl ester, 2-propenoic acid, 2-propenyl ester, unii-705ga5o3us, ccris 4745, acrylic acid allyl ester, 2-propenoic acid, 2-propen-1-yl ester, allylacrylate, acrylic acid allyl, acmc-20alut
C=CCOC(=O)C=C
112.128
112.13
Technical

Chemical Identifiers

999-55-3
112.128
QTECDUFMBMSHKR-UHFFFAOYSA-N
13835
C=CCOC(=O)C=C
C6H8O2
MFCD00014949
allyl acrylate, acrylic acid, allyl ester, 2-propenoic acid, 2-propenyl ester, unii-705ga5o3us, ccris 4745, acrylic acid allyl ester, 2-propenoic acid, 2-propen-1-yl ester, allylacrylate, acrylic acid allyl, acmc-20alut
prop-2-enyl prop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H310-H330-H226-H302-H315-H319-H335
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

RTECSNumber : UD3675000

TSCA : TSCA

Documents