missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Allyl acrylate, 95%, stab. with 4-methoxyphenol

Manufacturer:  Alfa Aesar™ 4446314

 View more versions of this product

Catalog No. AA4446314



Specifications

Specifications

Allyl acrylate
32°C (89°F)
1.436
25g
999-55-3
MFCD00014949
1743576
Slightly soluble in water.
C=CCOC(=O)C=C
112.128
112.13
95%
0.935 g/mL
Pungent
46°C to 47°C (40mmHg)
Stabilized with 4-methoxyphenol
C6H8O2
UN2929
2-propenoic acid, 2-propen-1-yl ester
QTECDUFMBMSHKR-UHFFFAOYSA-N
prop-2-enyl prop-2-enoate
13835
Liquid

Chemical Identifiers

999-55-3
112.128
QTECDUFMBMSHKR-UHFFFAOYSA-N
13835
C=CCOC(=O)C=C
C6H8O2
MFCD00014949
2-propenoic acid, 2-propen-1-yl ester
prop-2-enyl prop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H310-H330-H226-H302-H315-H319-H335
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H311-H315-H319-H330-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

EINECSNumber : 213-662-2

RTECSNumber : UD3675000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents